Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridyl Ethyl Ketone, MP Biomedicals

Catalog No. ICN220569 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220569 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220569 Supplier MP Biomedicals, Inc Supplier No. 0522056910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3238-55-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
InChI Key ZHAZHKPVEROFLH-UHFFFAOYSA-N
Synonym 2-propionylpyridine, 1-pyridin-2-yl propan-1-one, 1-propanone, 1-2-pyridinyl, 2-pyridyl ethyl ketone, 1-2-pyridinyl-1-propanone, 1-propanone, 1-2-pyridyl, 2-propanylpyridine, methylacetylpyridine, 2-propionyl pyridine, ethyl-2-pyridylketone
PubChem CID 520574
IUPAC Name 1-pyridin-2-ylpropan-1-one
SMILES CCC(=O)C1=CC=CC=N1

Specifications

Quantity 10 g
Chemical Name or Material 2-Pyridyl Ethyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.