CAS RN 32398-69-9
IUPAC Name:
hept-4-yn-3-ol
Synonyms:
hept-4-yn-3-ol
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
FMOKAMXPKGORIR-UHFFFAOYNA-N
SMILES:
CCC#CC(O)CC
1
–
1
of
1
results
1
4-Heptyn-3-ol, 98%, Thermo Scientific™
CAS: 32398-69-9 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.172 MDL Number: MFCD00041592 InChI Key: FMOKAMXPKGORIR-UHFFFAOYSA-N Synonym: 4-heptyn-3-ol,3-heptyn-5-ol,acmc-1aifi PubChem CID: 141704 IUPAC Name: hept-4-yn-3-ol SMILES: CCC#CC(CC)O