CAS RN 32431-84-8
IUPAC Name:
3-fluorothiophene-2-carboxylic acid
Synonyms:
3-fluorothiophene-2-carboxylic acid
Molecular Weight (g/mol):
146.14
Molecular Formula:
C5H3FO2S
InChi Key:
WPHRBUAOSDHRDS-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C=CS1
1
–
1
of
1
results
1
3-Fluoro-2-thiophenecarboxylic Acid 98.0+%, TCI America™
CAS: 32431-84-8 Molecular Formula: C5H3FO2S Molecular Weight (g/mol): 146.14 MDL Number: MFCD03426911 InChI Key: WPHRBUAOSDHRDS-UHFFFAOYSA-N PubChem CID: 819050 IUPAC Name: 3-fluorothiophene-2-carboxylic acid SMILES: OC(=O)C1=C(F)C=CS1