Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-2-thiophenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F08501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08501G Supplier TCI America Supplier No. F08501G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32431-84-8
Molecular Formula C5H3FO2S
Molecular Weight (g/mol) 146.14
MDL Number MFCD03426911
InChI Key WPHRBUAOSDHRDS-UHFFFAOYSA-N
PubChem CID 819050
IUPAC Name 3-fluorothiophene-2-carboxylic acid
SMILES OC(=O)C1=C(F)C=CS1

Specifications

Melting Point 177°C
Color White
Quantity 1 g
Formula Weight 146.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-2-thiophenecarboxylic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.