accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3244-88-0

IUPAC Name: disodium 2-amino-5-[(4-amino-3-sulfophenyl)[(1Z)-4-imino-3-methyl-5-sulfonatocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Synonyms: acid fuchsin
Molecular Weight (g/mol): 585.53
Molecular Formula: C20H17N3Na2O9S3
InChi Key: WZRZTHMJPHPAMU-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC1=C\C(C=C(C1=N)S([O-])(=O)=O)=C(\C1=CC=C(N)C(=C1)S(O)(=O)=O)C1=CC=C(N)C(=C1)S([O-])(=O)=O
Quantity 
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
Color 
  • (6)
Grade 
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (2)
  • (3)
  • (8)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (5)
  • (3)
  • (1)