Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fuchsin Acid Staining Solution, 1%
SCI_ED

Catalog No. S25694 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25694 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25694
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Fuchsin Acid Staining Solution, 1%
CAS 3244-88-0, 7732-18-5
Assay Percent Range 1% w/v
Packaging 1%
Density ∽1.00g/mL
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.