accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32444-63-6

H3CNCH3N(E)

CAS RN 32444-63-6

IUPAC Name: (2E)-3-[4-(dimethylamino)phenyl]prop-2-enenitrile
Synonyms: (2E)-3-[4-(dimethylamino)phenyl]prop-2-enenitrile
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: CSCOHKCYDZVRMQ-ONEGZZNKSA-N
SMILES: CN(C)C1=CC=C(\C=C\C#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results