Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Dimethylaminocinnamonitrile 98.0+%, TCI America™
SDP

Catalog No. D13375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13375G Supplier TCI America Supplier No. D13375G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32444-63-6
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD00001933
InChI Key CSCOHKCYDZVRMQ-ONEGZZNKSA-N
Synonym trans-4-dimethylaminocinnamonitrile, 3-4-dimethylamino phenyl acrylonitrile, e-3-4-dimethylaminophenyl prop-2-enenitrile, 2e-3-4-dimethylamino phenyl prop-2-enenitrile, e-3-4-dimethylamino phenyl acrylonitrile, p-dimethylamino cinnamonitrile, cinnamonitrile, p-dimethylamino, 2-propenenitrile, 3-4-dimethylamino phenyl, 4-dimethylamino cinnamonitrile
PubChem CID 5374281
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enenitrile
SMILES CN(C)C1=CC=C(C=C1)C=CC#N

Specifications

Melting Point 166°C
Color Yellow
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Dimethylaminocinnamonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.