accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CNS

CAS RN 32446-47-2

IUPAC Name: 3-ethyl-1,3-benzothiazol-3-ium bromide
Synonyms: 3-ethyl-1,3-benzothiazol-3-ium bromide
Molecular Weight (g/mol): 244.15
Molecular Formula: C9H10BrNS
InChi Key: XBWYYTURPBHPFJ-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1=CSC2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)