Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethylbenzothiazolium Bromide 98.0+%, TCI America™
SDP

Catalog No. E039510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E039510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E039510G Supplier TCI America Supplier No. E039510G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32446-47-2
Molecular Formula C9H10BrNS
Molecular Weight (g/mol) 244.15
MDL Number MFCD00051852
InChI Key XBWYYTURPBHPFJ-UHFFFAOYSA-M
Synonym 3-ethylbenzothiazolium bromide, benzothiazolium, 3-ethyl-, bromide 1:1, benzothiazolium, 3-ethyl-, bromide, 3-ethyl-1,3-benzothiazol-3-ium bromide, 3-ethylbenzothiazoliumbromide, acmc-209ht4, 3-ethylbenzo d thiazol-3-ium bromide
PubChem CID 182081
IUPAC Name 3-ethyl-1,3-benzothiazol-3-ium bromide
SMILES [Br-].CC[N+]1=CSC2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 244.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethylbenzothiazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.