accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 324575-10-2

IUPAC Name: tributyl(methyl)phosphanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; tributyl(methyl)phosphanium
Molecular Weight (g/mol): 497.49
Molecular Formula: C15H30F6NO4PS2
InChi Key: YJPDLBMZLGTDRZ-UHFFFAOYSA-N
SMILES: CCCC[P+](C)(CCCC)CCCC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results