Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylmethylphosphonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. T26805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26805G Supplier TCI America Supplier No. T26805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 324575-10-2
Molecular Formula C15H30F6NO4PS2
Molecular Weight (g/mol) 497.492
InChI Key YJPDLBMZLGTDRZ-UHFFFAOYSA-N
PubChem CID 44630020
IUPAC Name bis(trifluoromethylsulfonyl)azanide;tributyl(methyl)phosphanium
SMILES CCCC[P+](C)(CCCC)CCCC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 497.49
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tributylmethylphosphonium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.