accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32556-71-1

IUPAC Name: oct-1-yn-3-ol
Synonyms: 1-octyn-3-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: VUGRNZHKYVHZSN-UHFFFAOYNA-N
SMILES: CCCCCC(O)C#C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results