Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-Octyn-3-ol 99.0+%, TCI America™
SDP

Catalog No. O02355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02355G 5 g
O02351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02355G Supplier TCI America Supplier No. O02355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32556-71-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00191475
InChI Key VUGRNZHKYVHZSN-MRVPVSSYSA-N
Synonym s-1-octyn-3-ol, s---1-octyn-3-ol, s-3-hydroxy-1-octyne, 1-octyn-3-ol, 3s, 3s-1-octyn-3-ol, unii-d8ho63xx92, 3s-oct-1-yn-3-ol, 3s-hydroxy-1-octyne, --1-octyn-3-ol, 1-octyn-3-ol, s
PubChem CID 2734534
IUPAC Name (3S)-oct-1-yn-3-ol
SMILES CCCCCC(C#C)O

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 126.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-1-Octyn-3-ol [omega Side-Chain Unit for PG Synthesis]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.