accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3257-18-9

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-(benzylsulfanyl)propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-(benzylsulfanyl)propanoic acid
Molecular Weight (g/mol): 345.41
Molecular Formula: C18H19NO4S
InChi Key: ATPNWHGYKFXQNF-UHFFFAOYNA-N
SMILES: OC(=O)C(CSCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results