Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-S-benzyl-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. C26145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26145G 5 g
C261425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26145G Supplier TCI America Supplier No. C26145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3257-18-9
Molecular Formula C18H19NO4S
Molecular Weight (g/mol) 345.413
MDL Number MFCD00004785
InChI Key ATPNWHGYKFXQNF-INIZCTEOSA-N
Synonym N-Cbz-S-benzyl-L-cysteine, Z-Cys(Bzl)-OH
PubChem CID 98775
IUPAC Name (2R)-3-benzylsulfanyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CSCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 345.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-S-benzyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.