accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3260-85-3

IUPAC Name: 4-chloro-1-methoxy-2-methylbenzene
Synonyms: 4-chloro-1-methoxy-2-methylbenzene
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: VDYDAUQHTVCCBX-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1C
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)