Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxytoluene 97.0+%, TCI America™
SDP

Catalog No. C17245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17245G Supplier TCI America Supplier No. C17245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3260-85-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00070771
InChI Key VDYDAUQHTVCCBX-UHFFFAOYSA-N
Synonym 4-chloro-2-methylanisole, 5-chloro-2-methoxytoluene, unii-7g3kq2a26k, 2-methyl-4-chloroanisole, labotest-bb ltbb003018, labotest-bb lt03497723, 4-chloro-1-methoxy-2-methyl-benzene, 4-chloro-2-methyl anisole, pubchem2665, benzene, 4-chloro-1-methoxy-2-methyl
PubChem CID 76748
IUPAC Name 4-chloro-1-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)OC

Specifications

Color White
Quantity 5 g
Formula Weight 156.61
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.