accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32607-23-1

IUPAC Name: dilanthanum(3+) tris(2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate))
Synonyms: dilanthanum(3+) tris(2,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,4-bis(olate))
Molecular Weight (g/mol): 898.7
Molecular Formula: C18Cl6La2O12
InChi Key: XFCYTTPBGABCBW-UHFFFAOYSA-H
SMILES: [La+3].[La+3].[O-]C1=C(Cl)C(=O)C([O-])=C(Cl)C1=O.[O-]C1=C(Cl)C(=O)C([O-])=C(Cl)C1=O.[O-]C1=C(Cl)C(=O)C([O-])=C(Cl)C1=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)