Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Chloranilate, MP Biomedicals

Catalog No. MP520823901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520823901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP520823901 Supplier MP Biomedicals, Inc Supplier No. 0520823901
Add to Cart
Add to Cart

Reagent grade chemical for biomedical research

  • Important Notice: This product belongs to MP Biomedicals Rare Chemical library
  • Most Rare Chemical products are available for immediate delivery and sold 'As Is'
  • MP Biomedicals does not collect analytical data for those products
  • However, analytical test may be performed upon custom request for bulk orders

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32607-23-1
Molecular Formula C18H6Cl6La2O12
Molecular Weight (g/mol) 904.745
MDL Number MFCD03939079
InChI Key GGKWUCDKAJOTCW-UHFFFAOYSA-N
Synonym chloranilic acid lanthanum salt, chloranilic acid, lanthanum salt
PubChem CID 126959841
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;lanthanum
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.[La].[La]

Specifications

Quantity 1 g
Linear Formula C18O12Cl6La2
Formula Weight 898.78
Grade Reagent
Chemical Name or Material LANTHANUM CHLORANILATE
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.