CAS RN 32619-42-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oleuropein, ≥98% (HPLC), MilliporeSigma™ Supelco™
Oleuropein is a phenolic compound with pungent odor, which is found in extra virgin olive oil. It can undergo hydrolysis by Lactobacillus plantarum via action of β-glucosidase and esterase to form elenolic acid and hydroxytyrosol as the final products.