Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleuropein, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013930 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3930 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3930 Supplier MilliporeSigma Supelco Supplier No. 9216710MG
Add to Cart
Add to Cart

Oleuropein is a phenolic compound with pungent odor, which is found in extra virgin olive oil. It can undergo hydrolysis by Lactobacillus plantarum via action of β-glucosidase and esterase to form elenolic acid and hydroxytyrosol as the final products.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32619-42-4
Physical Form Neat
Linear Formula C25H32O13
MDL Number MFCD16879008
Synonym (2 S,3 E,4 S)-3-Ethylidene-2-(beta-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2 H-pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester
Type Analytical Standard
Quantity 10 mg
Formula Weight 540.51
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.