accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32634-66-5

IUPAC Name: (2R,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Synonyms: (2R,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular Weight (g/mol): 386.36
Molecular Formula: C20H18O8
InChi Key: CMIBUZBMZCBCAT-IYBDPMFKSA-N
SMILES: CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)