accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32638-88-3

IUPAC Name: 2-(4,5,6-trihydroxy-3-oxo-3H-xanthen-9-yl)benzene-1-sulfonate
Synonyms: 2-(3,4,5-trihydroxy-6-oxoxanthen-9-yl)benzenesulfonate
Molecular Weight (g/mol): 399.35
Molecular Formula: C19H11O8S
InChi Key: JTZKNOUMMWIARI-UHFFFAOYSA-M
SMILES: OC1=CC=C2C(OC3=C(O)C(=O)C=CC3=C2C2=CC=CC=C2S([O-])(=O)=O)=C1O
Physical Form 
  • (2)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results