accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3PCH3CH3H3C

CAS RN 32673-25-9

IUPAC Name: di-tert-butyl(phenyl)phosphane
Synonyms: di-tert-butyl(phenyl)phosphane
Molecular Weight (g/mol): 222.31
Molecular Formula: C14H23P
InChi Key: XOJNEFQLMRCOMS-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)