Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Tert-Butylphenylphosphine

Catalog No. 6824115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-241-15G 5 g
68-241-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 68-241-15G Supplier Sigma Aldrich Supplier No. 6824115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Cross-coupling reactionsPalladium-catalyzed monoarylation reactions (cocatalyst)Coupling of alkyl bromides with aryl boronic acidsReactant for the synthesis of palladium-FiberCats for aryl chloride Suzuki coupling

Specifications

CAS 32673-25-9
Density 0.933 g/mL at 25°C
Boiling Point 124°C to 128°C (10 mmHg)
Molecular Formula C14H23P
Refractive Index n20/D 1.5382
Linear Formula C14H23P
MDL Number MFCD00049096
Quantity 5 g
Synonym NSC 244300
Molecular Weight (g/mol) 222.31 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.