accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOHOOHOOHOOH

CAS RN 32701-19-2

IUPAC Name: 3-({2-[(2-carboxyethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)propanoic acid
Synonyms: 3-({2-[(2-carboxyethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)propanoic acid
Molecular Weight (g/mol): 320.3
Molecular Formula: C12H20N2O8
InChi Key: IZQWNOQOAFSHOK-UHFFFAOYSA-N
SMILES: OC(=O)CCN(CCN(CCC(O)=O)CC(O)=O)CC(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results