Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine-N,N'-diacetic-N,N'-dipropionic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. E02881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02881G Supplier TCI America Supplier No. E02881G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity crystalline powder for laboratory research applications.

Chemical Identifiers

CAS 32701-19-2
Molecular Formula C12H20N2O8
Molecular Weight (g/mol) 320.298
MDL Number MFCD00002768
InChI Key IZQWNOQOAFSHOK-UHFFFAOYSA-N
Synonym EDDADP
PubChem CID 544630
IUPAC Name 3-[2-[2-carboxyethyl(carboxymethyl)amino]ethyl-(carboxymethyl)amino]propanoic acid
SMILES C(CN(CCN(CCC(=O)O)CC(=O)O)CC(=O)O)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 320.30
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine-N,N′-diacetic-N,N′-dipropionic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.