accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32703-80-3

CH3H3CCH3NN

CAS RN 32703-80-3

IUPAC Name: 4-tert-butylbenzene-1,2-dicarbonitrile
Synonyms: 4-tert-butylbenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: LOTMIRVNJTVTSU-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C#N)C(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results