Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphthalonitrile 98.0+%, TCI America™
SDP

Catalog No. B13345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13345G 5 g
B133425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13345G Supplier TCI America Supplier No. B13345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32703-80-3
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00060112
InChI Key LOTMIRVNJTVTSU-UHFFFAOYSA-N
Synonym 4-tert-Butyl-1,2-dicyanobenzene
PubChem CID 122931
IUPAC Name 4-tert-butylbenzene-1,2-dicarbonitrile
SMILES CC(C)(C)C1=CC(=C(C=C1)C#N)C#N

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphthalonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.