accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32705-91-2

IUPAC Name: 3-({2-[(2-carboxyethyl)amino]ethyl}amino)propanoic acid dihydrochloride
Synonyms: 3-({2-[(2-carboxyethyl)amino]ethyl}amino)propanoic acid dihydrochloride
Molecular Weight (g/mol): 277.14
Molecular Formula: C8H18Cl2N2O4
InChi Key: GSRDLTQJRCGIQI-UHFFFAOYSA-N
SMILES: Cl.Cl.OC(=O)CCNCCNCCC(O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)