Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine-N,N'-dipropionic Acid Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. E023825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E023825G 25 g
E02381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E023825G Supplier TCI America Supplier No. E023825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 32705-91-2 | C8H18Cl2N2O4 | 277.14 g/mol

Chemical Identifiers

CAS 32705-91-2
Molecular Formula C8H18Cl2N2O4
Molecular Weight (g/mol) 277.14
MDL Number MFCD00060168
InChI Key GSRDLTQJRCGIQI-UHFFFAOYSA-N

Specifications

Melting Point 225°C
Color White
Quantity 25 g
Formula Weight 277.14
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine-N,N′-dipropionic Acid Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.