accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32737-35-2

CH3H3CH3CCH3OHOHOOHOHO

CAS RN 32737-35-2

IUPAC Name: 4,4,4',4'-tetramethyl-3,3',4,4'-tetrahydro-2,2'-spirobi[[1]benzopyran]-6,6',7,7'-tetrol
Synonyms: 4,4,4',4'-tetramethyl-3H,3'H-2,2'-spirobi[[1]benzopyran]-6,6',7,7'-tetrol
Molecular Weight (g/mol): 372.42
Molecular Formula: C21H24O6
InChi Key: SZUGPQFFJXSTDD-UHFFFAOYSA-N
SMILES: CC1(C)CC2(CC(C)(C)C3=CC(O)=C(O)C=C3O2)OC2=CC(O)=C(O)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results