Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6',7,7'-Tetrahydroxy-4,4,4',4'-tetramethyl-2,2'-spirobichroman 98.0+%, TCI America™
SDP

Catalog No. T17915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17915G Supplier TCI America Supplier No. T17915G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32737-35-2
Molecular Formula C21H24O6
Molecular Weight (g/mol) 372.42
MDL Number MFCD00627043
InChI Key SZUGPQFFJXSTDD-UHFFFAOYSA-N
Synonym 4,4,4′C,4′C-Tetramethyl-2,2′C-spirobi[chroman]-6,6′C,7,7′C-tetraol
PubChem CID 350598
IUPAC Name 4,4,4',4'-tetramethyl-3,3',4,4'-tetrahydro-2,2'-spirobi[[1]benzopyran]-6,6',7,7'-tetrol
SMILES CC1(C)CC2(CC(C)(C)C3=CC(O)=C(O)C=C3O2)OC2=CC(O)=C(O)C=C12

Specifications

Quantity 5 g
Formula Weight 372.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′,7,7′-Tetrahydroxy-4,4,4′,4′-tetramethyl-2,2′-spirobichroman
EINECSNumber (5)-5650
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.