accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3275-24-9

IUPAC Name: titanium(4+) tetrakis(dimethylazanide)
Synonyms: tetrakis(dimethylamido)titanium
Molecular Weight (g/mol): 224.18
Molecular Formula: C8H24N4Ti
InChi Key: MNWRORMXBIWXCI-UHFFFAOYSA-N
SMILES: [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (4)
  • (2)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (5)
  • (4)
  • (1)