Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrakis(dimethylamido)titanium(IV)

Catalog No. 4698585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-985-85G 5 g
46-985-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-985-85G Supplier Sigma Aldrich Supplier No. 4698585G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Please inquire for bulk quantity, pricing, and packaging options.Tetrakis(dimethylamido)titanium(IV) (TDMAT) is a precursor to titanium nitride (TiN) thin films by organometallic chemical vapor deposition (OMCVD)and titanium dioxide thin films by atomic layer deposition (ALD).TDMAT undergoes exothermal reaction with excess cyclopentadiene to yield tris(dimethylamido)(++5-cyclopentadienyl)titanium(IV).

Specifications

CAS 3275-24-9
Density 0.947 g/mL (at 25°C)
Boiling Point 50°C (0.5 mmHg)
Molecular Formula C8H24N4Ti
Linear Formula [(CH3)2 N]4 Ti
MDL Number MFCD00014861
Quantity 5 g
Synonym TDMAT; Tetrakis(dimethylamino)titanium(IV)
Molecular Weight (g/mol) 224.17
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.