accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHNBr

CAS RN 32774-29-1

IUPAC Name: 2-(5-bromo-1H-indol-3-yl)ethan-1-ol
Synonyms: 2-(5-bromo-1H-indol-3-yl)ethanol
Molecular Weight (g/mol): 240.1
Molecular Formula: C10H10BrNO
InChi Key: ZENXDUDCTZLSRP-UHFFFAOYSA-N
SMILES: OCCC1=CNC2=CC=C(Br)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results