Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoindole-3-ethanol 98.0+%, TCI America™
SDP

Catalog No. B5068200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5068200MG 200 mg
B50681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5068200MG Supplier TCI America Supplier No. B5068200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32774-29-1
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00130167
InChI Key ZENXDUDCTZLSRP-UHFFFAOYSA-N
Synonym 5-Bromo-3-(2-hydroxyethyl)indole
PubChem CID 122956
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanol
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCO

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 200 mg
Formula Weight 240.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.