accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNClI

CAS RN 32779-38-7

IUPAC Name: 2-chloro-5-iodopyrimidine
Synonyms: 2-chloro-5-iodopyrimidine
Molecular Weight (g/mol): 240.43
Molecular Formula: C4H2ClIN2
InChi Key: WSZRCNZXKKTLQE-UHFFFAOYSA-N
SMILES: ClC1=NC=C(I)C=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)