Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-iodopyrimidine, 95%

Catalog No. AAH63423MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63423MD 250 mg
AAH6342303 1 g
AAH6342306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63423MD Supplier Thermo Scientific Chemicals Supplier No. H63423MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 32779-38-7 | C4H2ClIN2 | 240.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32779-38-7
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.43
MDL Number MFCD06200210
InChI Key WSZRCNZXKKTLQE-UHFFFAOYSA-N
PubChem CID 1714223
IUPAC Name 2-chloro-5-iodopyrimidine
SMILES ClC1=NC=C(I)C=N1

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Synonym pyrimidine, 2-chloro-5-iodo, 2-chloro-5-iodo-pyrimidine, pubchem21488, acmc-209hvo, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, ksc569e7p, pyrimidine,2-chloro-5-iodo
Formula Weight 240.43
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-iodopyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.