accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 3278-82-8

IUPAC Name: 1,5-dibromoanthracene
Synonyms: 1,5-dibromoanthracene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: DIMYVOCPPKNNPF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=CC3=C(Br)C=CC=C3C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results