Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dibromoanthracene 97.0+%, TCI America™
SDP

Catalog No. D31831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31831G Supplier TCI America Supplier No. D31831G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3278-82-8
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.026
InChI Key DIMYVOCPPKNNPF-UHFFFAOYSA-N
PubChem CID 14923878
IUPAC Name 1,5-dibromoanthracene
SMILES C1=CC2=CC3=C(C=CC=C3Br)C=C2C(=C1)Br

Specifications

Melting Point 209°C
Color Yellow
Quantity 1 g
Formula Weight 336.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dibromoanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.