CAS RN 3279-27-4
IUPAC Name:
2-(2-methylbutan-2-yl)phenol
Synonyms:
2-(2-methylbutan-2-yl)phenol
Molecular Weight (g/mol):
164.25
Molecular Formula:
C11H16O
InChi Key:
BGRKGHSKCFAPCL-UHFFFAOYSA-N
SMILES:
CCC(C)(C)C1=CC=CC=C1O
1
–
1
of
1
results
1
2-tert-Amylphenol 98.0+%, TCI America™
CAS: 3279-27-4 Molecular Formula: C11H16O Molecular Weight (g/mol): 164.248 InChI Key: BGRKGHSKCFAPCL-UHFFFAOYSA-N Synonym: 2-(1,1-Dimethylpropyl)phenol, 2-tert-Pentylphenol PubChem CID: 76769 IUPAC Name: 2-(2-methylbutan-2-yl)phenol SMILES: CCC(C)(C)C1=CC=CC=C1O