Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Amylphenol 98.0+%, TCI America™
SDP

Catalog No. A188525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A188525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A188525G Supplier TCI America Supplier No. A188525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3279-27-4
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
InChI Key BGRKGHSKCFAPCL-UHFFFAOYSA-N
Synonym 2-(1,1-Dimethylpropyl)phenol, 2-tert-Pentylphenol
PubChem CID 76769
IUPAC Name 2-(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 g
UN Number 3145
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-tert-Amylphenol
EINECSNumber (3)-0503
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.