accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 328-80-3

IUPAC Name: 3-nitro-5-(trifluoromethyl)benzoate
Synonyms: 3-nitro-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 234.11
Molecular Formula: C8H3F3NO4
InChi Key: ODCLHXGXGFBBTA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=CC(=C1)C(F)(F)F)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results