Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Trifluoromethyl-5-nitrobenzoic Acid, TCI America™
SDP

Catalog No. T13445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13445G Supplier TCI America Supplier No. T13445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 328-80-3
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
MDL Number MFCD00024509
InChI Key ODCLHXGXGFBBTA-UHFFFAOYSA-N
Synonym 3-nitro-5-trifluoromethyl benzoic acid, 3-nitro-5-trifluoromethylbenzoic acid, 3-trifluoromethyl-5-nitrobenzoic acid, 3-carboxy-5-nitrobenzotrifluoride, 3-nitro-5 trifluoromethyl benzoic acid, benzoic acid, 3-nitro-5-trifluoromethyl, pubchem4544, acmc-1cr4t, rarechem al bo 1012
PubChem CID 616602
SMILES C1=C(C=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 235.12
Physical Form Crystalline Powder
Chemical Name or Material 3-Trifluoromethyl-5-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.