accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClFFF

CAS RN 328-91-6

IUPAC Name: 2-chloro-1,4-bis(trifluoromethyl)benzene
Synonyms: 2-chloro-1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 248.55
Molecular Formula: C8H3ClF6
InChi Key: IDNVTCBYGYOPKJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results