accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32804-77-6

IUPAC Name: 2-ethoxyethyl 2-cyanoacetate
Synonyms: 2-ethoxyethyl 2-cyanoacetate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: UMCLCZMPTREESK-UHFFFAOYSA-N
SMILES: CCOCCOC(=O)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results