CAS RN 328073-92-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ VEGF Inducer, GS4012
A cell-permeable pyridinyl-thioether compound that acts as a potent inducer of VEGF and VEGF-mediated vessel formation.