Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ VEGF Inducer, GS4012

Catalog No. 67649110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-649-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-649-110MG Supplier MilliporeSigma™ Supplier No. 67649110MG
Add to Cart
Add to Cart

A cell-permeable pyridinyl-thioether compound that acts as a potent inducer of VEGF and VEGF-mediated vessel formation.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins Potent inducer of VEGF and VEGF-mediated vessel formation
CAS 328073-92-3
Molecular Formula C14H15NOS·HCl
Formula Weight 281.8g/mol
Solubility DMSO, water, or MeOH
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.